Learn More
Cayman Chemical Piperaquine, Purity: ≥98%, CAS: 4085-31-8, 50 mg
SDP
Catalog No. 502982270
Encompass_Preferred
An antimalarial agent; a bis-quinoline compound; inhibits the ex vivo growth of P. falciparum isolates from malaria-infected patients (IC50s = 11.8-217.3 nM)
Catalog No. 50-298-2270 Supplier Cayman Chemical Supplier No. 2079950 MG
An antimalarial agent; a bis-quinoline compound; inhibits the ex vivo growth of P. falciparum isolates from malaria-infected patients (IC50s = 11.8-217.3 nM)