Learn More
Enzo Life Sciences PPH . hydrochloride (10mg)
SDP
Supplier:  Enzo Life Sciences ALX430080M010
Encompass_Preferred
Effective, non-cell permeable and non-toxic analog of CPH for the detection of superoxide radicals. Resistant against reduction of vitamin C. For cell permeable analog see CPH (ALX-430-078). Alternative name: 1-Hydroxy-4-phosphono-oxy-2,2,6,6-tetramethylpiperidine HCl. Purity: ≥99% (EPR). Appearance: White to off-white solid. Solubility: Soluble in DMSO, 100% ethanol, methanol or water. Long Term Storage: -20°C.
Catalog No. 50-200-9189