Learn More
Medchemexpress LLC PPI-2458 | 431077-35-9 | 99.7% | 424.53 | 1 ML
SDP
Catalog No. 5000350394
Encompass_Preferred
PPI-2458 is a potent, orally active, selective, and irreversible inhibitor of methionine aminopeptidase-2 (MetAP-2), primarily used in arthritis and lymphoma research.
  • Potent, orally active, selective, and irreversible MetAP-2 inhibitor.
  • Inhibits HUVEC proliferation with a GI50 of 0.2 nM.
  • Inhibits proliferation and MetAP-2 in SU-DHL-16 cells.
  • Reverses joint swelling and inflammation in PG-PS-induced arthritis models.
  • Suppresses tumor growth in severe combined immunodeficient mice with SR tumor xenografts.
Catalog No. 50-003-50394 Supplier Medchemexpress LLC Supplier No. HY1373110MM/1ML
PPI-2458 is a potent, orally active, selective, and irreversible inhibitor of methionine aminopeptidase-2 (MetAP-2), primarily used in arthritis and lymphoma research.
  • Potent, orally active, selective, and irreversible MetAP-2 inhibitor.
  • Inhibits HUVEC proliferation with a GI50 of 0.2 nM.
  • Inhibits proliferation and MetAP-2 in SU-DHL-16 cells.
  • Reverses joint swelling and inflammation in PG-PS-induced arthritis models.
  • Suppresses tumor growth in severe combined immunodeficient mice with SR tumor xenografts.