Learn More
Medchemexpress LLC PPI-2458 | 431077-35-9 | 99.7% | 424.53 | C22H36N2O6 | 5 MG
SDP
Catalog No. 5000222421
Encompass_Preferred
PPI-2458 is a potent, orally active, selective and irreversible inhibitor of methionine aminopeptidase-2 (MetAP-2). A member of the fumagillin class, it is used in preclinical research to evaluate antiangiogenic and antiproliferative effects in models of rheumatoid arthritis and lymphoma. The compound is supplied as a powder with defined storage recommendations and is available in milligram-scale pack sizes suitable for laboratory studies.
  • Selective, irreversible inhibition of MetAP-2.
  • Orally active in preclinical models.
  • High chemical purity suitable for research.
  • Supplied in milligram pack sizes for lab use.
  • Recommended storage: powder -20°C (long term) or 4°C (short term).
Catalog No. 50-002-22421 Supplier Medchemexpress LLC Supplier No. HY137315MG
PPI-2458 is a potent, orally active, selective and irreversible inhibitor of methionine aminopeptidase-2 (MetAP-2). A member of the fumagillin class, it is used in preclinical research to evaluate antiangiogenic and antiproliferative effects in models of rheumatoid arthritis and lymphoma. The compound is supplied as a powder with defined storage recommendations and is available in milligram-scale pack sizes suitable for laboratory studies.
  • Selective, irreversible inhibition of MetAP-2.
  • Orally active in preclinical models.
  • High chemical purity suitable for research.
  • Supplied in milligram pack sizes for lab use.
  • Recommended storage: powder -20°C (long term) or 4°C (short term).