Learn More
Medchemexpress LLC Ppi-2458 | 431077-35-9 | 99.7% | C22H36N2O6 | 10MG
SDP
Catalog No. 5000259105
Encompass_Preferred
PPI-2458 is a small-molecule, orally active, selective and irreversible inhibitor of methionine aminopeptidase-2 (MetAP-2). It is a fumagillin-class derivative used in preclinical research to study angiogenesis, cell proliferation, and for arthritis and lymphoma models. The compound is supplied as a solid with high reported purity for research use.
  • Potent, selective MetAP-2 inhibition suitable for mechanistic studies.
  • Orally active in preclinical models.
  • Irreversible binding provides sustained target engagement.
  • High chemical purity (~99.7%) supports reproducible results.
  • Stable solid with defined storage conditions for extended shelf life.
Catalog No. 50-002-59105 Supplier Medchemexpress LLC Supplier No. HY1373110MG
PPI-2458 is a small-molecule, orally active, selective and irreversible inhibitor of methionine aminopeptidase-2 (MetAP-2). It is a fumagillin-class derivative used in preclinical research to study angiogenesis, cell proliferation, and for arthritis and lymphoma models. The compound is supplied as a solid with high reported purity for research use.
  • Potent, selective MetAP-2 inhibition suitable for mechanistic studies.
  • Orally active in preclinical models.
  • Irreversible binding provides sustained target engagement.
  • High chemical purity (~99.7%) supports reproducible results.
  • Stable solid with defined storage conditions for extended shelf life.