Learn More
Aviva Systems Biology Corp PPIA Antibody - middle region (ARP51352_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51352_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIA. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PPIA. Alternate names: CYPA, CYPH, MGC117158, MGC12404, MGC23397ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-60556