Learn More
TARGETMOL CHEMICALS INC PPIASE-PARVULIN INHIBITO 5MG
SDP
Catalog No. 502580977
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PPIase-Parvulin Inhibitor is a cell-pemeable inhibitor of the Pin1 and Pin4. purity: 98%
Catalog No. 50-258-0977 Supplier TARGETMOL CHEMICALS INC Supplier No. T221355MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PPIase-Parvulin Inhibitor is a cell-pemeable inhibitor of the Pin1 and Pin4. purity: 98%