Learn More
Cayman Chemical PPIase-ParvulIn InhIbItr 10mg
SDP
Catalog No. 502967835
Encompass_Preferred
A cell-permeable inhibitor of the PPIases Pin1 and Pin4 (IC50s = 1.5 and 1.0 µM, respectively), interacting with the PPIase domain; blocks the proliferation of cancer cells that overexpress Pin1 and Pin4 (IC50 range = 2-5 µM), and similarly inhibits wild MEFs but not Pin1-/- MEFs
Catalog No. 50-296-7835 Supplier Cayman Chemical Supplier No. 1721410 MG
Add to Cart
Add to Cart
A cell-permeable inhibitor of the PPIases Pin1 and Pin4 (IC50s = 1.5 and 1.0 µM, respectively), interacting with the PPIase domain; blocks the proliferation of cancer cells that overexpress Pin1 and Pin4 (IC50 range = 2-5 µM), and similarly inhibits wild MEFs but not Pin1-/- MEFs