Learn More
Cayman Chemical PPIase-Parvulin Inhibitor, Purity: ≥95%, CAS: 64005-90-9, 5 mg
SDP
Catalog No. 502966819
Encompass_Preferred
A cell-permeable inhibitor of the PPIases Pin1 and Pin4 (IC50s = 1.5 and 1.0 µM, respectively), interacting with the PPIase domain; blocks the proliferation of cancer cells that overexpress Pin1 and Pin4 (IC50 range = 2-5 µM), and similarly inhibits wild MEFs but not Pin1-/- MEFs
Catalog No. 50-296-6819 Supplier Cayman Chemical Supplier No. 172145 MG
A cell-permeable inhibitor of the PPIases Pin1 and Pin4 (IC50s = 1.5 and 1.0 µM, respectively), interacting with the PPIase domain; blocks the proliferation of cancer cells that overexpress Pin1 and Pin4 (IC50 range = 2-5 µM), and similarly inhibits wild MEFs but not Pin1-/- MEFs