Learn More
Aviva Systems Biology Corp Ppic Antibody - C-terminal region (ARP44369_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44369_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ppic. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Ppic. Alternate names: CyP-20cApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-140