Learn More
Aviva Systems Biology Corp PPID Antibody - N-terminal region (ARP56624_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56624_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPID. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPID. Alternate names: CYP-40, CYPD, MGC33096ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61929