Learn More
Aviva Systems Biology Corp PPIE Antibody - middle region (ARP40626_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40626_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIE. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPIE. Alternate names: CYP33, CYP-33ApplicationWB. Species reactivity: Guinea Pig, Human, Rat, Yeast, Zebrafish
Catalog No. 50-014-57555