Learn More
Aviva Systems Biology Corp PPIE Antibody - middle region (ARP40627_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40627_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIE. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PPIE. Alternate names: CYP33, CYP-33ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-014-57556