Learn More
Aviva Systems Biology Corp PPIF Antibody - middle region (ARP46141_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46141_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIF. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPIF. Alternate names: CYP3, Cyp-D, FLJ90798, MGC117207ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-014-53461