Learn More
Aviva Systems Biology Corp PPIG Antibody - C-terminal region (ARP65692_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65692_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIG. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human PPIG. Alternate names: CARS-Cyp, CYP, MGC133241, SRCyp, SCAF10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-541