Learn More
Aviva Systems Biology Corp PPIH Antibody - middle region (ARP52154_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52154_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPIH. Alternate names: CYP-20, CYPH, MGC5016, SnuCyp-20, USA-CYPApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-285-652