Learn More
Aviva Systems Biology Corp PPIL2 Antibody - C-terminal region (ARP55026_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55026_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PPIL2. Alternate names: CYC4, CYP60, FLJ39930, MGC33174, MGC787, hCyP-60, Cyp-60ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61401