Learn More
Aviva Systems Biology Corp PPIL3 Antibody - middle region (ARP51174_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51174_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPIL3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPIL3. Alternate names: CYPJApplicationWB. Species reactivity: Human
Catalog No. 50-014-60510