Learn More
ABclonal Technology PPIL4 Rabbit pAb
SDP
Catalog No. 5000104469
Encompass_Preferred
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.
Catalog No. 50-001-04469 Supplier ABclonal Technology Supplier No. A15920500UL
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.