Learn More
Aviva Systems Biology Corp PPM1A Antibody - middle region (ARP57765_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57765_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPM1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPM1A. Alternate names: MGC9201, PP2C-ALPHA, PP2CA, PP2CalphaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62151