Learn More
Aviva Systems Biology Corp PPM1B Antibody - N-terminal region (ARP57767_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPM1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPM1B. Alternate names: MGC21657, PP2C-beta-X, PP2CB, PP2CBETA, PPC2BETAXApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62152