Learn More
Aviva Systems Biology Corp PPM1J Antibody - C-terminal region (ARP54745_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54745_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPM1J. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PPM1J. Alternate names: DKFZp434P1514, FLJ35951, MGC19531, MGC90149, PP2CZ, PP2Czeta, PPP2CZ, RP11-426L16.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61299