Learn More
Aviva Systems Biology Corp PPM1M Antibody - middle region (ARP52710_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52710_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPM1M. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPM1M. Alternate names: FLJ32332, PP2C-eta, PP2CE, PP2CetaApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-759