Learn More
Sino Biological Recombinant Mouse KDM4B/JMJD2B Protein (GST Tag)
Catalog No. 502574507
Encompass_Preferred
Recombinant mouse KDM4B (1-725) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-4507 Supplier Sino Biological Supplier No. K42431BG100
Recombinant mouse KDM4B (1-725) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.