Learn More
Aviva Systems Biology Corp PPP1CB Antibody - C-terminal region (ARP65811_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65811_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1CB. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human PPP1CB. Alternate names: MGC3672, PP-1B, PP1beta, PPP1CDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-288-576