Learn More
Aviva Systems Biology Corp PPP1CC Antibody - C-terminal region (ARP64668_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64668_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1CC. It was validated on Western Blot. . Alternate names: PP1gamma, PPP1GApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-330