Learn More
Aviva Systems Biology Corp PPP1R13B Antibody - middle region (ARP47979_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47979_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R13B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPP1R13B. Alternate names: ASPP1, KIAA0771, p53BP2-like, p85ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59694