Learn More
Aviva Systems Biology Corp PPP1R13L Antibody - middle region (ARP32826_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32826_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R13L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPP1R13L. Alternate names: IASPP, NKIP1, RAI, iASPP gene, RAI4ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54779