Learn More
Aviva Systems Biology Corp PPP1R14A Antibody - N-terminal region (ARP63944_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63944_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R14A. It was validated on Western Blot. . Alternate names: CPI-17, CPI17, PPP1INLApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-288-127