Learn More
Aviva Systems Biology Corp PPP1R16B Antibody - N-terminal region (ARP62226_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62226_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R16B. It was validated on Western Blot. . Alternate names: ANKRD4, KIAA0823, TIMAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-620