Learn More
Aviva Systems Biology Corp Ppp1r1a Antibody - N-terminal region (ARP42314_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42314_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ppp1r1a. It was validated on Western Blot. . Alternate names: 0610038N18Rik, I-1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-992