Learn More
Aviva Systems Biology Corp PPP1R2 Antibody - N-terminal region (ARP61382_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61382_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R2. It was validated on Western Blot. . Alternate names: IPP2, MGC87148, IPP-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-359