Learn More
Aviva Systems Biology Corp PPP1R3A Antibody - N-terminal region (ARP45349_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45349_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPP1R3A. Alternate names: PP1G, PPP1R3, GMApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59010