Learn More
Aviva Systems Biology Corp PPP1R3C Antibody - N-terminal region (ARP62902_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62902_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R3C. It was validated on Western Blot. . Alternate names: PPP1R5ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-287-800