Learn More
Aviva Systems Biology Corp PPP1R8 Antibody - middle region (ARP40412_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40412_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R8. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human PPP1R8. Alternate names: ARD1, ARD-1, NIPP1, NIPP-1, PRO2047ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53196