Learn More
Aviva Systems Biology Corp PPP1R8 Antibody - middle region (ARP40411_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40411_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R8. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPP1R8. Alternate names: ARD-1, ARD1, NIPP-1, NIPP1, PRO2047ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-54058