Learn More
Aviva Systems Biology Corp PPP1R8 Antibody - N-terminal region (ARP40754_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40754_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP1R8. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human PPP1R8. Alternate names: ARD-1, ARD1, NIPP-1, NIPP1, PRO2047ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-889