Learn More
Aviva Systems Biology Corp PPP2CA Antibody - N-terminal region (ARP63057_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63057_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP2CA. It was validated on Western Blot. . Alternate names: PP2Ac, PP2CA, PP2Calpha, RP-CApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-857