Learn More
Aviva Systems Biology Corp Ppp2r2c Antibody - N-terminal region (ARP57784_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57784_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ppp2r2c. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 6330548O06Rik, IMYPNO, IMYPNO1, PR52ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-376