Learn More
Aviva Systems Biology Corp Ppp2r2d Antibody - C-terminal region (ARP56157_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56157_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ppp2r2d. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1300017E19Rik, D7Ertd753e, MDS026ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-121