Learn More
Aviva Systems Biology Corp PPP2R3B Antibody - C-terminal region (ARP42370_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42370_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP2R3B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human PPP2R3B. Alternate names: NY-REN-8, PPP2R3L, PPP2R3LY, PR48, NYREN8ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58203