Learn More
Aviva Systems Biology Corp PTPA Antibody - N-terminal region (ARP62067_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62067_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP2R4. It was validated on Western Blot. . Alternate names: MGC2184, PP2A, PR53, PTPAApplicationWB. Species reactivity: Human
Catalog No. 50-287-556