Learn More
Aviva Systems Biology Corp PPP2R5A Antibody - N-terminal region (ARP48033_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48033_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP2R5A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPP2R5A. Alternate names: B56A, MGC131915, PR61AApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59706