Learn More
Aviva Systems Biology Corp PPP2R5C Antibody - middle region (ARP57771_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57771_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP2R5C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPP2R5C. Alternate names: B56G, MGC23064, PR61GApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62154