Learn More
Aviva Systems Biology Corp Ppp2r5e Antibody - N-terminal region (ARP56694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ppp2r5e. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 4633401M22Rik, AI449017, B56betaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-196