Learn More
Aviva Systems Biology Corp Ppp3cb Antibody - C-terminal region (ARP57516_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57516_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ppp3cb. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1110063J16Rik, Calnb, CnAbeta, CnabApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-328