Learn More
Aviva Systems Biology Corp PPP3R1 Antibody - N-terminal region (ARP56124_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56124_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP3R1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PPP3R1. Alternate names: CALNB1, CNB, CNB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61728