Learn More
Aviva Systems Biology Corp PPP4C Antibody - middle region (ARP58516_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58516_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP4C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PPP4C. Alternate names: PP4, PPH3, PPX, PP4C, PPP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62337