Learn More
ABclonal Technology PPP4C Rabbit pAb
SDP
Catalog No. 5000097431
Encompass_Preferred
Enables protein serine/threonine phosphatase activity. Involved in regulation of double-strand break repair via homologous recombination. Located in cytosol, nucleoplasm, and plasma membrane.
Catalog No. 50-000-97431 Supplier ABclonal Technology Supplier No. A13531100UL
Enables protein serine/threonine phosphatase activity. Involved in regulation of double-strand break repair via homologous recombination. Located in cytosol, nucleoplasm, and plasma membrane.