Learn More
Aviva Systems Biology Corp PPP4R1 Antibody - middle region (ARP62616_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62616_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PPP4R1. It was validated on Western Blot. . Alternate names: PP4(Rmeg), PP4R1ApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-287-724